EMAIL THIS PAGE TO A FRIEND
Journal of cellular biochemistry
Role of ROS and MAPK in TPA-induced ICAM-1 expression in the myeloid ML-1 cell line.
PMID 17131377
Abstract
Intercellular adhesion molecule 1 (ICAM-1) has been implicated in playing a key role in the mechanism of inflammatory process initiated in response to environmental agents, and during normal hematopoietic cell differentiation. Though induction of ICAM-1 by 12-O-tetradecanoyl-phorbol-13-acetate (TPA) in myeloid cells has been reported, the molecular mechanism by which TPA upregulates ICAM-1 expression remains unclear. In the present study, we investigated the signaling mechanism associated with TPA-induced ICAM-1 expression in ML-1 cells. Herein, our microarray, flow cytometry, and Western blot analysis indicated that ICAM-1 was constitutively expressed at a low level in ML-1 cells, but its expression was further upregulated at both the mRNA and protein levels in response to TPA. ICAM-1 expression in response to TPA was inhibited by pretreatment with GF109203X [a specific inhibitor of protein kinase C (PKC)], or with PD98059 and U0126 (specific inhibitors of MEK), suggesting the importance of PKC, and Erk1/2 signaling cascades in this response. Interestingly, ICAM-1 expression in response to TPA-induced PKC activation was linked to the generation of reactive oxygen species (ROS), as pretreatment with NAC (an ROS scavenger) blocked both ErK1/2 activation and ICAM-1 expression induced by TPA. In addition, TPA-induced ICAM-1 expression was blocked by inhibition of nuclear factor-kappaB (NF-kappaB) activation following pretreatment with BAY11-7085 (a specific inhibitor of NF-kappaB activation). TPA-induced NF-kappaB activation was shown by increased degradation of IkB (NF-kappaB specific inhibitory protein). Together, these observations demonstrated that TPA, a potent activator of PKC, induces ICAM-1 expression via a ROS- and ERK1/2-dependent signaling mechanism in ML-1 cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
ML-1, 88113007