EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Lithium diisopropylamide solvated by hexamethylphosphoramide: substrate-dependent mechanisms for dehydrobrominations.
PMID 17132006
Abstract
Lithium diisopropylamide-mediated dehydrobrominations of exo-2-bromonorbornane, 1-bromocyclooctene, and cis-4-bromo-tert-butylcyclohexane were studied in THF solutions and THF solutions with added hexamethylphosphoramide (HMPA). Rate studies reveal a diverse array of mechanisms based on mono-, di-, and trisolvated monomers as well as triple ions. The results are contrasted with analogous eliminations in THF in the absence of HMPA.