EMAIL THIS PAGE TO A FRIEND
Hellenic journal of cardiology : HJC = Hellenike kardiologike epitheorese
Should atenolol still be recommended as first-line therapy for primary hypertension?
PMID 17134065