EMAIL THIS PAGE TO A FRIEND
Organic letters
Nonenzymatic oligomerization of RNA by TNA templates.
PMID 17134278
Abstract
Cytosine TNA promotes nonenzymatic, template-directed oligomerization of complementary activated rGMP, leading to selective and efficient formation of RNA products. This process models "genetic takeover" of a pre-RNA by RNA. [reaction: see text]
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T7642
L-(+)-Threose, ≥60%, syrup
C4H8O4