EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Carbolithiation of diphenylacetylene as a stereoselective route to (Z)-tamoxifen and related tetrasubstituted olefins.
PMID 17137396
Abstract
Carbolithiation of diphenylacetylene can be exploited to generate (E)-1-lithio-1,2-diphenylalkyl-1-enes which can be reacted in situ with triisopropylborate to stereoselectively provide (E)-1,2-diphenyl-1-alkylene boronic acids. These tetrasubstituted vinylboronic acids served as versatile intermediates for the generation of tetrasubstituted olefins with retention of stereochemistry. The application of this method for the stereoselective synthesis of (Z)-tamoxifen and related analogues is described.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D204803
Diphenylacetylene, 98%
C14H10