EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Determination of esomeprazole and its two main metabolites in human, rat and dog plasma by liquid chromatography with tandem mass spectrometry.
PMID 17142113
Abstract
A LC-MS/MS method was developed for quantitative determination of esomeprazole, and its two main metabolites 5-hydroxyesomeprazole and omeprazole sulphone in 25 microL human, rat or dog plasma. The analytes and their internal standards were extracted from plasma into methyl tert-butyl ether - dichloromethane (3:2, v/v). After evaporation and reconstitution of the organic extract the analytes were separated on a reversed-phase LC column and measured by atmospheric-pressure positive ionisation MS. The linearity range was 20-20,000 nmol/L for esomeprazole and omeprazole sulphone, and 20-4000 nmol/L for 5-hydroxyesomeprazole. The extraction recoveries ranged between 80 and 105%. The intra- and inter-day imprecision were less than 9.5% with accuracy between 97.7% and 100.1% for all analytes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O0151000
Omeprazole impurity D, European Pharmacopoeia (EP) Reference Standard
C17H19N3O4S