EMAIL THIS PAGE TO A FRIEND
Chembiochem : a European journal of chemical biology
A gene cluster encoding rhizoxin biosynthesis in "Burkholderia rhizoxina", the bacterial endosymbiont of the fungus Rhizopus microsporus.
PMID 17154220