EMAIL THIS PAGE TO A FRIEND
Journal of thrombosis and haemostasis : JTH
Clinical sample investigation (CSI) hematology: pinpointing the precise onset of heparin-induced thrombocytopenia (HIT).
PMID 17155944
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E9251 Ecteola Cellulose, coarse