EMAIL THIS PAGE TO A FRIEND
Journal of microbiological methods
Detection of Tsh protein mucinolytic activity by SDS-PAGE.
PMID 17157399
Abstract
The temperature-sensitive hemagglutinin (Tsh, 140 kDa) produced by Escherichia coli is cleaved into a fragment (106 kDa) containing mucinase activity, and an agglutinin fragment (33 kDa). By incorporating mucins into SDS-PAGE gels stained by Schiff's periodic acid, we could simultaneously detect about 0.5 microg of mucinase activity and the fragment molecular mass.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H1136
Hyaluronidase from Streptomyces hyalurolyticus, lyophilized powder