EMAIL THIS PAGE TO A FRIEND
Neuro endocrinology letters
Extra- and intracellular formation of reactive oxygen species by human neutrophils in the presence of pheniramine, chlorpheniramine and brompheniramine.
PMID 17159800
Abstract
Allergic inflammation was found to be accompanied by activation of neutrophils. Since this resulted in increased formation of reactive oxygen species, the antioxidative activity of antiallergic drugs was considered to decrease the risk of tissue damage. However, if the drug-induced inhibition of radical formation occurred intracellularly, it could disturb regulation of neutrophil functions and decrease bactericidal activity of these cells. Separate analysis of extra- and intracellular activity of antioxidative drugs is thus of particular importance. To differentiate the effects of three antiallergic H1-antihistamine drugs (pheniramine, chlor- and brompheniramine) on radical formation outside and inside human neutrophils. Formation of reactive oxygen species was determined in vitro using the chemiluminescence method. The chemiluminescence signal, initiated by phorbol-12-myristate-13-acetate, was enhanced either with isoluminol (extracellular) or with luminol in the presence of extracellular scavengers, superoxide dismutase and catalase (intracellular). The antihistamines tested displayed dual activity - they inhibited the extracellular and potentiated the intracellular chemiluminescence. Chlor- and brompheniramine were found to be more effective than pheniramine. Compared to other H1-antihistamines (such as dithiaden or loratadine, active both extra- and intracellularly), the observed inhibition caused by the pheniramines tested was unique since it occurred selectively outside neutrophils. This might indicate the ability of these drugs to minimise toxic effects of extracellular radicals without affecting intracellular oxidant production involved in regulation of neutrophil functions and in microbial killing.