EMAIL THIS PAGE TO A FRIEND
Biotechnic & histochemistry : official publication of the Biological Stain Commission
A multichromatic stain for Lowicryl K4M embedded tissues.
PMID 1716158
Abstract
A staining procedure using celestine blue and chromotrope 2R for Lowicryl K4M embedded tissues is presented. The stain produces a reliable multichromatic stain for light microscopy on Lowicryl embedded semithin sections.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
206342
Celestine blue, Dye content 80 %
C17H18ClN3O4
C3143
Chromotrope 2R, suitable for modified Gomori Trichrome stain
C16H10N2Na2O8S2