EMAIL THIS PAGE TO A FRIEND
Nano letters
Single-pair FRET characterization of DNA tweezers.
PMID 17163711
Abstract
The self-assembly properties of DNA make it an ideal choice for the construction of nanomachines such as DNA tweezers. Because nanomachines function as individual units, they need to be studied on the single-molecule level. From single-pair FRET investigations, we show that "open" tweezers exist in a single conformation with minimal FRET efficiency, whereas upon addition of a "closing strand", three conformations are observed, which are averaged out in ensemble measurements.