EMAIL THIS PAGE TO A FRIEND
FEMS microbiology letters
Isolation and characterization of a new bacterium capable of biotransforming cis -epoxysuccinic acid to D(-) -tartaric acid.
PMID 17166227
Abstract
A bacterial strain 1-3 capable of enantioselectively hydrolyzing cis-epoxysuccinic acid to D(-)-tartaric acid was isolated from soil. Phenotypic characteristics, Biolog GN test and phylogenetic analysis of strain 1-3 indicated that it belongs to the genus Bordetella. cis-Epoxysuccinic acid could be used as a carbon source and inducer for production of cis-epoxysuccinic acid hydrolase. After the purification of the enzymatic biotransformed product, a colorless and scentless crystal was obtained, which was confirmed to be the tartaric acid with laevorotatory optical rotation by the (1)H nuclear magnetic resonance spectrum and optical rotation analysis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
757071
(+/-)-trans-Oxirane-2,3-dicarboxylic acid, 97%
C4H4O5