EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
RNA interference silencing of the adaptor molecules ShcC and Fe65 differentially affect amyloid precursor protein processing and Abeta generation.
PMID 17170108
Abstract
The amyloid precursor protein (APP) and its pathogenic by-product amyloid-beta protein (Abeta) play central roles in Alzheimer disease (AD) neuropathogenesis. APP can be cleaved by beta-secretase (BACE) and alpha-secretase to produce APP-C99 and APP-C83. These C-terminal fragments can then be cleaved by gamma-secretase to produce Abeta and p3, respectively. p3 has been reported to promote apoptosis, and Abeta is the key component of senile plaques in AD brain. APP adaptor proteins with phosphotyrosine-binding domains, including ShcA (SHC1), ShcC (SHC3), and Fe65 (APBB1), can bind to and interact with the conserved YENPTY motif in the APP-C terminus. Here we have described for the first time the effects of RNA interference (RNAi) silencing of ShcA, ShcC, and Fe65 expression on APP processing and Abeta production. RNAi silencing of ShcC led to reductions in the levels of APP-C-terminal fragments (APP-CTFs) and Abeta in H4 human neuroglioma cells stably overexpressing full-length APP (H4-FL-APP cells) but not in those expressing APP-C99 (H4-APP-C99 cells). RNAi silencing of ShcC also led to reductions in BACE levels in H4-FL-APP cells. In contrast, RNAi silencing of the homologue ShcA had no effect on APP processing or Abeta levels. RNAi silencing of Fe65 increased APP-CTF levels, although also decreasing Abeta levels in H4-FL-APP cells. These findings suggest that pharmacologically blocking interaction of APP with ShcC and Fe65 may provide novel therapeutic strategies against AD.