EMAIL THIS PAGE TO A FRIEND
Yeast (Chichester, England)
The ORF YNL274c (GOR1) codes for glyoxylate reductase in Saccharomyces cerevisiae.
PMID 17173333
Abstract
The enzyme glyoxylate reductase reversibly reduces glyoxylate to glycolate, or alternatively hydroxypyruvate to D-glycerate, using either NADPH or NADH as a co-factor. The enzyme has multiple metabolic roles in different organisms. In this paper we show that GOR1 (ORF YNL274c) encodes a glyoxylate reductase and not a hydroxyisocaproate dehydrogenase in Saccharomyces cerevisiae, even though it also has minor activity on alpha-ketoisocaproate. In addition, we show that deletion of the glyoxylate reductase-encoding gene leads to higher biomass concentration after diauxic shift.