EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Identification and kinetic study of tyrosinase inhibitors found in sake lees.
PMID 17177508
Abstract
The present study found that the n-hexane extract of freeze-dried sake lees inhibits tyrosinase activity and showed that the constituents isolated from the n-hexane extract are the mixture of triacylglycerols. The inhibitory effects of triolein and trilinolein found as the triacylglycerols were examined using tyrosinases from mushroom and Streptomyces castaneoglobisporus. The IC50 values of the triacylglycerol mixture for the oxidase activity on mushroom and Streptomyces tyrosinases were 20 and 0.14 microg/mL, respectively. The IC50 values of trilinolein for the oxidase activity on mushroom and Streptomyces tyrosinases were 8.4 and 0.1 microM, respectively. However, the inhibitory effect of triolein (IC50=30 microM) was lower than that of trilinolein, even when the Streptomyces tyrosinase was used for the assay. Kinetic analyses indicate that both trilinolein and triolein inhibit the tyrosinase activity noncompetitively. When transformed with a plasmid carrying the Streptomyces tyrosinase gene, the melanin-synthesizing ability of the transformed Escherichia coli host was dose-dependently interfered with by trilinolein.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T9517
Glyceryl trilinoleate, ≥98% (TLC), liquid
C57H98O6