EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Electrochemical synthesis of cobalt hydroxide films with tunable interlayer spacings.
PMID 17180232
Abstract
An electrochemical synthetic condition is developed to produce cobalt hydroxide films with significantly increased basal spacings (d(001)>or= 25.0 A) by incorporating anionic surfactants (i.e. sodium dodecyl sulfate and 1-hexadecanesulfonate) into the interlayer regions via electrodeposition.