EMAIL THIS PAGE TO A FRIEND
The New England journal of medicine
Carbapenems for surgical prophylaxis?
PMID 17182996
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A5737 Cefotetan disodium
C17H15N7Na2O8S4