EMAIL THIS PAGE TO A FRIEND
International journal of dermatology
Superficial cutaneous Trichosporon asahii infection in an immunocompetent host.
PMID 17184246
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
03907
Tioconazole, 97%
C16H13Cl3N2OS