EMAIL THIS PAGE TO A FRIEND
Molecular endocrinology (Baltimore, Md.)
Ubiquitination and proteasome-mediated degradation of BRCA1 and BARD1 during steroidogenesis in human ovarian granulosa cells.
PMID 17185394
Abstract
Germ-line mutations in BRCA1 predispose women to early-onset, familial breast and ovarian cancers. However, BRCA1 expression is not restricted to breast and ovarian epithelial cells. For example, ovarian BRCA1 expression is enriched in ovarian granulosa cells, which are responsible for ovarian estrogen production in premenopausal women. Furthermore, recent tissue culture and animal studies suggest a functional role of BRCA1 in ovarian granulosa cells. Although levels of BRCA1 are known to fluctuate significantly during folliculogenesis and steroidogenesis, the mechanism by which BRCA1 expression is regulated in granulosa cells remains to be elucidated. Here we show that the ubiquitin-proteasome degradation pathway plays a significant role in the coordinated protein stability of BRCA1 and its partner BARD1 in ovarian granulosa cells. Our work identifies the amino-terminal RING domain-containing region of BRCA1 as the degron sequence that is both necessary and sufficient for polyubiquitination and proteasome-mediated protein degradation. Interestingly, mutations in the RING domain that abolish the ubiquitin E3 ligase activity of BRCA1 do not affect its own ubiquitination or degradation in ovarian granulosa cells. The proteasome-mediated degradation of BRCA1 and BARD1 also occurs during the cAMP-dependent steroidogenic process. Thus, the dynamic changes of BRCA1/BARD1 protein stability in ovarian granulosa cells provide an excellent paradigm for investigating the regulation of this protein complex under physiological conditions.