EMAIL THIS PAGE TO A FRIEND
Chemistry & biodiversity
Ternary zinc(II)-dipeptide complexes for the hydrolytic cleavage of DNA at physiological pH.
PMID 17192010
Abstract
A series of Zn(II) complexes with cysteinylglycine (CysGly) and histidylserine (HisSer), and of CysGly and histidylphenylalanine (HisPhe) were investigated. Complex stabilities were determined potentiometrically, and binding geometries were probed by means of 1H-NMR spectroscopy, using Co(II) instead of Zn(II) as a spectroscopic marker. The ternary 1:1:1 complexes [Zn(II)(CysGly)(HisSer)] and [Zn(II)(CysGly)(HisPhe)] were shown by UV experiments, fluorescence titration, and gel electrophoresis to intercalate with DNA, and to hydrolytically cleave supercoiled DNA (form-I), partly also circular (form-II) DNA, under physiological conditions (37 degrees, H2O, pH 7.5).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H2504
His-Leu
C12H20N4O3
H3129
His-Ser
C9H14N4O4