EMAIL THIS PAGE TO A FRIEND
Chemistry & biodiversity
The studies of density, apparent molar volume, and viscosity of bovine serum albumin, egg albumin, and lysozyme in aqueous and RbI, CsI, and DTAB aqueous solutions at 303.15 K.
PMID 17192025
Abstract
Density (rho), apparent molar volume (V(phi)), and viscosity (eta) of 0.0010 to 0.0018% (w/v) of bovine serum albumin (BSA), egg albumin, and lysozyme in 0.0002, 0.0004, and 0.0008 M aqueous RbI and CsI, and (dodecyl)(trimethyl)ammonium bromide (DTAB) solutions were obtained. The experimental data were regressed against composition, and constants are used to elucidate the conformational changes in protein molecules. With salt concentration, the density of proteins is found to decrease, and the order of the effect of additives on density is observed as CsI > RbI > DTAB. The trend of apparent molar volume of proteins is found as BSA > egg-albumin > lysozyme for three additives. In general, eta values of BSA remain higher for all compositions of RbI than that of egg-albumin for CsI and DTAB. These orders of the data indicate the strength of intermolecular forces between proteins and salts, and are helpful for understanding the denaturation of proteins.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
251445
Rubidium iodide, 99.9% trace metals basis
IRb