EMAIL THIS PAGE TO A FRIEND
Organic letters
An efficient synthesis of epoxydiynes and a key fragment of neocarzinostatin chromophore.
PMID 17192081
Abstract
[reaction: see text] A key structural feature of the Neocarzinostatin chromophore is a reactive epoxydiyne. We present here a new method for the preparation of epoxydiynes by the addition of an allenyl zinc bromide to a propargylic ketone.