EMAIL THIS PAGE TO A FRIEND
Small (Weinheim an der Bergstrasse, Germany)
In vitro transcription and protein translation from carbon nanotube-DNA assemblies.
PMID 17193110
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
401633
1-Pyrenemethylamine hydrochloride, 95%
C17H13N · HCl