EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Spectral image analysis for the absorption bands of the beta-methallyl free radical in the vapor phase.
PMID 17204450
Abstract
The radicals formed in the flash photolysis of 2-methylbut-1-ene and subsequent reactions have been investigated by kinetic spectroscopy and gas liquid chromatography. Less than 10% of photo products are formed by a molecular made of fission of the excited olefin, and of the radical modes the relative probabilities of band fission, beta(CH):beta(CH):alpha(CC) are 13:1.37:1. The extinction coefficients of beta-methallyl radical measured experimentally for all the absorption bands. The decay of the beta-methallyl radical was second order. The rate constant for the beta-methallyl radical recombination experimentally measured was 2.6+/-0.3 x 10(10) l mol(-1)s(-1) at 295+/-2K. The spectrum image showing the absorption bands was examined by image processing techniques in order to improve the visual experience of each band by localizing to a specific region of interest. Experimental results illustrate how the exact location of absorption bands was clearly extracted from the spectral image and further improvements in the visual detection of absorption bands.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
295469
2-Methylpropene, 99%
C4H8