EMAIL THIS PAGE TO A FRIEND
Supportive care in cancer : official journal of the Multinational Association of Supportive Care in Cancer
A meta-analysis comparing the efficacy of four 5-HT3-receptor antagonists for acute chemotherapy-induced emesis.
PMID 17205281
Abstract
Comparing antiemetic efficacy of different 5-HT(3)-receptor antagonists (5-HT(3)RAs) is difficult due to inter-study variability. Therefore, a meta-analysis was performed to comparatively evaluate dolasetron, granisetron, ondansetron and tropisetron for acute chemotherapy-induced nausea and vomiting (CINV). Comparisons between 5-HT(3)RAs were based on 44 randomized studies (including 12,343 patients) identified by MEDLINE, CANCERLIT or EMBASE searches and subcategorized by chemotherapy type (cisplatin- or non-cisplatin-based). When all studies were combined, granisetron was equivalent to ondansetron (n = 27), and showed an advantage vs tropisetron (p = 0.018; n = 12). Ondansetron vs tropisetron (n = 11) and ondansetron vs dolasetron (n = 3) revealed equivalence in each comparison. An advantage for 3 mg granisetron vs 8 mg ondansetron was found in non-cisplatin-based studies (p = 0.015; n = 6). Overall equivalence was seen between ondansetron, 24 or 32 mg, and granisetron, 2 or 3 mg, for all studies (n = 13). There was a possible advantage for higher (24 or 32 mg) vs lower (8 mg) ondansetron dose regimens with cisplatin-based trials (n = 6). No differences were seen between 3 and 1 mg granisetron doses (n = 6). Efficacy of 5-HT(3)RAs for preventing CINV following cisplatin- and non-cisplatin-based chemotherapy is comparable, with the exception of granisetron vs tropisetron. Some differences were noted in dosing subanalyses.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9571
Dolasetron mesylate hydrate, ≥98% (HPLC), powder
C19H20N2O3 · CH4O3S · xH2O