EMAIL THIS PAGE TO A FRIEND
Artificial Organs
Electrically conductive biodegradable polymer composite for nerve regeneration: electricity-stimulated neurite outgrowth and axon regeneration.
PMID 17209956