EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Silibinin suppresses PMA-induced MMP-9 expression by blocking the AP-1 activation via MAPK signaling pathways in MCF-7 human breast carcinoma cells.
PMID 17214970
Abstract
Matrix metalloproteinase-9 (MMP-9) plays an important role in the invasion and metastasis of cancer cells. In this study, we examined the inhibitory effect of silibinin, a flavonoid antioxidant from milk thistle (Silybum marianum L.) on PMA-induced MMP-9 expression in MCF-7 human breast carcinoma cells. Silibinin significantly and selectively suppressed PMA-induced MMP-9 expression in MCF-7. Silibinin has been found to inhibit PMA-induced MMP-9 gene transcriptional activity by blocking the activation of AP-1 via MAPK signaling pathways. Moreover, the Matrigel invasion assay showed that silibinin reduces PMA-induced invasion of MCF-7 cells. These results suggest that silibinin represents a potential anti-metastatic agent suppressing PMA-induced cancer cell invasion through the specific inhibition of AP-1-dependent MMP-9 gene expression.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0104 Ascochlorin, ≥98% (HPLC), from Verticillium hemipterigenum
C23H29ClO4