EMAIL THIS PAGE TO A FRIEND
Organic letters
Asymmetric synthesis of cyclopropylamines starting from N-sulfinyl alpha-chloro ketimines.
PMID 17217261
Abstract
Treatment of novel chiral N-sulfinyl alpha-chloro ketimines with Grignard reagents resulted in the synthesis of chiral N-(1-substituted cyclopropyl)-tert-butanesulfinamides in acceptable to good yields and diastereoselectivity via 1,3-dehydrohalogenation and subsequent addition of the Grignard reagent to the intermediate cyclopropylideneamine. Only in the case of allylmagnesium chloride did the reaction lead to aziridines in high yield. Further deprotection toward N-unprotected 1-substituted cyclopropylamines was established, and the absolute configuration was determined. [reaction: see text].
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
125504
Cyclopropylamine, 98%
C3H7N