EMAIL THIS PAGE TO A FRIEND
Organic letters
Synthesis of protected chiral vicinal diaminoalcohols by diastereoselective intramolecular benzylic substitution from bistrichloroacetimidates.
PMID 17217276
Abstract
An efficient synthesis of chiral dihydrooxazines (2) from 1-aryl-2-amino-propane-1,3-diols (1) via the corresponding bistrichloroacetimidate intermediates has been developed. In this transformation, one trichloroacetimidate acts as a leaving group and the other acts as a nucleophile. The cyclization proceeds through an SN1 mechanism to provide trans-dihydrooxazines with complete diastereoselectivity irrespective of the absolute configuration of the benzylic alcohol. The transformation of 2 into other selectively protected aminodiols is also documented. [reaction: see text].
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T53805
Trichloroacetonitrile, 98%
C2Cl3N