EMAIL THIS PAGE TO A FRIEND
Organic letters
Multicomponent synthesis of 3-indolepropionic acids.
PMID 17217290
Abstract
A three-component one-pot procedure (3-MC) was developed to assemble 3-indolepropionic acids from commercially available materials. This new methodology affords the title compounds in high yields and without the use of chromatography. [reaction: see text].
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
220027
3-Indolepropionic acid, ReagentPlus®, 99%
C11H11NO2