EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Removal of alachlor and pretilachlor by laboratory-synthesized zerovalent iron in pesticide formulation solution.
PMID 17219301
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31251
Pretilachlor, PESTANAL®, analytical standard
C17H26ClNO2