EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Effect of fungicide seed treatments on N2-fixation and nodulation in pea, Pisum sativum L.
PMID 17219311
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45371
Carboxine, PESTANAL®, analytical standard
C12H13NO2S