EMAIL THIS PAGE TO A FRIEND
Journal of biotechnology
Serine scanning: a tool to prove the consequences of N-glycosylation of proteins.
PMID 17224199
Abstract
N-Glycosylation of proteins is a common posttranslational modification in eukaryotes. Often this results in enhanced protein stability through protection by the attached sugar moieties. Due to its 13 potential N-glycosylation motifs (N-X-T/S), recombinant hydroxynitrile lyase isoenzyme 5 from almonds (PaHNL5) is secreted by the heterologous host Pichia pastoris in a massively glycosylated form, and it shows extraordinary stability at low pH. The importance of N-glycosylation in general, and individual glycosylation sites in particular for stability at low pH were investigated. To identify especially important glycosylation sites asparagine from all N-X-S/T-motifs was replaced by serine. Thus, critical sites, which contributed to overall enzyme activity and/or stability, were identified individually. One glycosylation site revealed to be essential for stability at low pH. After enzymatic deglycosylation, leaving only one acetylglucosamine attached to asparagines, PaHNL5 retained most of its stability at low pH. Protonation effects in the active site as well as higher-order aggregational events upon incubation in low pH were excluded. This study provides evidence for the interconnection of N-glycosylation and stability at low pH for PaHNL5. Moreover, serine scanning was proven to be applicable for quick identification of critical glycosylation sites.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
116025
Mandelonitrile, technical grade
C8H7NO