EMAIL THIS PAGE TO A FRIEND
Biomedical chromatography : BMC
Chiral separation of three agrochemical toxins enantiomers by high-performance liquid chromatography on a vancomycin crystalline degradation products-chiral stationary phase.
PMID 17230451
Abstract
This work aims to evaluate for the enantiomeric separations of three agrochemical toxins: haloxyfop-methyl, fenoxaprop-p-ethyl and indoxacarb on crystalline degradation products-chiral stationary phase (CDP-CSP) of high-performance liquid chromatography (HPLC) under normal and polar organic phases. In the normal phase, the mobile phase was n-hexane with alcohols including methanol and isopropanol as polar modifiers. In the polar organic phase mode, the mobile phase was methanol with different percentages of triethylammunium acetate. The influence of flow rate (0.3-0.9 mL/min), analyte concentration and silica gel particle sizes (10, 15 and 30 microm) was investigated. This new chiral stationary phase showed excellent stereoselectivity for the two enantiomers of haloxyfop-methyl and fenoxaprop-p-ethyl and chiral recognition for indoxacarb under normal-phase mode. However, under polar organic phase, only indoxacarb was separated (alpha < 1.5). The chromatographic results were compared with commercial chiral columns.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45518
Fenoxaprop-ethyl, PESTANAL®, analytical standard
C18H16ClNO5
45817
Haloxyfop, PESTANAL®, analytical standard
C15H11ClF3NO4