EMAIL THIS PAGE TO A FRIEND
Biomacromolecules
Lipase-catalyzed synthesis of an epoxy-functionalized polyester from the suberin monomer cis-9,10-epoxy-18-hydroxyoctadecanoic acid.
PMID 17243765
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
379700
10-Hydroxydecanoic acid, technical grade
C10H20O3