EMAIL THIS PAGE TO A FRIEND
FEBS letters
Deletion of a MFS transporter-like gene in Cercospora nicotianae reduces cercosporin toxin accumulation and fungal virulence.
PMID 17250832
Abstract
Many phytopathogenic Cercospora species produce a host-nonselective polyketide toxin, called cercosporin, whose toxicity exclusively relies on the generation of reactive oxygen species. Here, we describe a Cercospora nicotianae CTB4 gene that encodes a putative membrane transporter and provide genetic evidence to support its role in cercosporin accumulation. The predicted CTB4 polypeptide has 12 transmembrane segments with four conserved motifs and has considerable similarity to a wide range of transporters belonging to the major facilitator superfamily (MFS). Disruption of the CTB4 gene resulted in a mutant that displayed a drastic reduction of cercosporin production and accumulation of an unknown brown pigment. Cercosporin was detected largely from fungal hyphae of ctb4 disruptants, but not from the surrounding medium, suggesting that the mutants were defective in both cercosporin biosynthesis and secretion. Cercosporin purified from the ctb4 disruptants exhibited toxicity to tobacco suspension cells, insignificantly different from wild-type, whereas the disruptants formed fewer lesions on tobacco leaves. The ctb4 null mutants retained normal resistance to cercosporin and other singlet oxygen-generating photosensitizers, indistinguishable from the parental strain. Transformation of a functional CTB4 clone into a ctb4 null mutant fully revived cercosporin production. Thus, we propose that the CTB4 gene encodes a putative MFS transporter responsible for secretion and accumulation of cercosporin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C6696
Cercosporin from Cercospora hayii
C29H26O10