EMAIL THIS PAGE TO A FRIEND
Journal of natural products
Antifungal acylcyclopentenediones from fruiting bodies of Hygrophorus chrysodon.
PMID 17253868
Abstract
Chrysotriones A (1) and B (2), two new 2-acylcyclopentene-1,3-dione derivatives, were isolated from the fruiting bodies of the Basidiomycete Hygrophorus chrysodon, and their structures were established by spectroscopic data and synthesis of compound 2. They represent the first examples of 2-acylcyclopentene-1,3-diones found in mushrooms and suggest an unprecedented biosynthetic pathway, starting from fatty acid derivatives. Initial tests indicated interesting antifungal activity of chrysotriones against Fusarium verticillioides, one of the common worldwide pathogens of cultivated plants.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
161683
4-Cyclopentene-1,3-dione, 95%
C5H4O2
249432
Undecanoyl chloride, 99%
C11H21ClO