EMAIL THIS PAGE TO A FRIEND
Neuropharmacology
Identification of ginsenoside interaction sites in 5-HT3A receptors.
PMID 17257631
Abstract
We previously demonstrated that 20(S)-ginsenoside Rg(3) (Rg(3)), one of the active components of Panax ginseng, non-competitively inhibits 5-HT(3A) receptor channel activity on extracellular side of the cell. Here, we sought to elucidate the molecular mechanisms underlying Rg(3)-induced 5-HT(3A) receptor regulation. We used the two-microelectrode voltage-clamp technique to investigate the effect of Rg(3) on 5-HT-mediated ion currents (I(5-HT)) in Xenopus oocytes expressing wild-type or 5-HT(3A) receptors harboring mutations in the gating pore region of transmembrane domain 2 (TM2). In oocytes expressing wild-type 5-HT(3A) receptors, Rg(3) dose-dependently inhibited peak I(5-HT) with an IC(50) of 27.6+/-4.3microM. Mutations V291A, F292A, and I295A in TM2 greatly attenuated or abolished the Rg(3)-induced inhibition of peak I(5-HT). Mutation V291A but not F292A and I295A induced constitutively active ion currents with decrease of current decay rate. Rg(3) accelerated the rate of current decay with dose-dependent manner in the presence of 5-HT. Rg(3) and TMB-8, an open channel blocker, dose-dependently inhibited constitutively active ion currents. The IC(50) values of constitutively active ion currents in V291A mutant receptor were 72.4+/-23.1 and 6.5+/-0.7microM for Rg(3) and TMB-8, respectively. Diltiazem did not prevent Rg(3)-induced inhibition of constitutively active ion currents in occlusion experiments. These results indicate that Rg(3) inhibits 5-HT(3A) receptor channel activity through interactions with residues V291, F292, and I295 in the channel gating region of TM2 and further demonstrate that Rg(3) regulates 5-HT(3A) receptor channel activity in the open state at different site(s) from those of TMB-8 and diltiazem.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T111
8-(Diethylamino)octyl-3,4,5-trimethoxybenzoate hydrochloride, ≥98%
C22H37NO5 · HCl