EMAIL THIS PAGE TO A FRIEND
Nature Immunology
Epithelial cells trigger frontline immunoglobulin class switching through a pathway regulated by the inhibitor SLPI.
PMID 17259987
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
07-167
Anti-Blys/TALL-1/BAFF/THANK Antibody, clone, Upstate®, from rabbit