EMAIL THIS PAGE TO A FRIEND
Yao xue xue bao = Acta pharmaceutica Sinica
[A new indole alkaloid glycoside from stems of Nauclea officinalis].
PMID 17262948
Abstract
To investigate the chemical constituents of water-soluble part of stem of Nauclea officinalis. The compounds were isolated and purified by resins, silica gel, and Sephadex LH20 column chromatography repeatedly, and their structures were identified by spectral analysis. Seven compounds were isolated and identified as: naucleamide A-10-O-beta-D-glucopyranoside (I), 5-beta-carboxystrictosidin (II), sweroside (III), loganin (IV), 3,4-dimethoxyphenyl-beta-D-glucopyranoside (V), 3, 4, 5-trimethoxyphenyl-beta-D-glucopyranoside (VI), 2-phenethylrutinoside (VII). Compound I is a new indole alkaloid glucoside, compounds I-VII were isolated from this plant for the first time.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SMB00083
Sweroside, ≥95% (LC/MS-ELSD)
C16H22O9