EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
The first catalytic Sakurai reaction of N-alkoxycarbonylamino sulfones with allyltrimethylsilane.
PMID 17268636
Abstract
We report the first catalytic Sakurai reaction of N-alkoxycarbonylamino sulfones with allylsilanes. The allylation reaction of N-alkoxycarbonylamino phenylsulfones with allyltrimethylsilane proceeded smoothly with low catalyst loading of bismuth triflate (2.0 mol%) to afford the corresponding protected homoallylic amines in moderate to very good yields (up to 96%).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
208264
Allyltrimethylsilane, 98%
C6H14Si