EMAIL THIS PAGE TO A FRIEND
Schizophrenia research
High glycine levels are associated with prepulse inhibition deficits in chronic schizophrenia patients.
PMID 17276036
Abstract
Prepulse inhibition (PPI) of the startle reflex, a measure of sensorimotor gating, is decreased in schizophrenia. The validity of a glutamatergic, N-methyl-d-aspartate receptor (NMDAR)-mediated model of PPI disruption is presently equivocal. The NMDAR antagonist ketamine disrupts PPI in rodents, but may increase PPI in healthy volunteers. Glycine (GLY), which acts as an obligatory co-agonist at the NMDAR-GLY site, induces PPI deficits in rats although, consistent with the hypo-NMDAR hypothesis, improves negative and cognitive symptoms in schizophrenia patients. We assessed the hypothesis that GLY serum levels may affect PPI parameters in schizophrenia. Forty-five chronically ill medicated schizophrenia patients and 37 matched healthy comparison subjects were tested for PPI of the eyeblink component of the startle reflex measured by electromyogram recording. Patients' demographic variables, symptom severity scores and GLY, serine and glutamate serum levels were obtained. Patients showed deficient PPI in blocks two and three of the PPI session and differed from controls in terms of change of degree of PPI as a function of the prepulse to eliciting stimulus interval. GLY levels correlated negatively with PPI parameters, such that patients with the highest GLY levels showed decreased PPI (rs=-0.4, p=0.03). These preliminary findings indirectly support previous observations on ketamine effects upon PPI in humans and suggest a dissociation of symptomatology and PPI changes as function of NMDAR modulation in schizophrenia.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H50004
4-Hydroxyphenylacetic acid, 98%
C8H8O3
G1001
D-Glutamic acid, ≥99% (TLC)
C5H9NO4
67419 Glycine 1 M solution
A4159
L-Asparagine, BioReagent, suitable for cell culture, suitable for insect cell culture
C4H8N2O3
A0884
L-Asparagine, ≥98% (HPLC)
C4H8N2O3
51363
L-Asparagine, certified reference material, TraceCERT®
C4H8N2O3
PHR1104
L-Aspartic acid, Pharmaceutical Secondary Standard; Certified Reference Material
C4H7NO4
51572
L-Aspartic acid, certified reference material, TraceCERT®
C4H7NO4
G2150
L-Glutamine solution, 200 mM, Hybri-Max, sterile-filtered, suitable for hybridoma
39496
L-Methionine, certified reference material, TraceCERT®
C5H11NO2S
PHR1100
L-Phenylalanine, Pharmaceutical Secondary Standard; Certified Reference Material
C9H11NO2
40541
L-Phenylalanine, certified reference material, TraceCERT®
C9H11NO2
91331
L-Phenylalanine, analytical standard, for Nitrogen Determination According to Kjeldahl Method
C9H11NO2
PHR1103
L-Serine, Pharmaceutical Secondary Standard; Certified Reference Material
C3H7NO3
54763
L-Serine, certified reference material, TraceCERT®
C3H7NO3
59202C
L-Glutamine Solution 200 mM, 29.23 mg/mL in saline, solution, suitable for cell culture
00920
N-Acetyl-L-aspartic acid, ≥99.0% (T)
C6H9NO5
A5930
N-Acetyl-Asp-Glu, ≥97% (TLC), powder
C11H16N2O8