EMAIL THIS PAGE TO A FRIEND
Medical physics
Comment on: "Basic investigations on the performance of a normoxic polymer gel with tetrakis-hydroxy-methyl-phosphonium chloride as an oxygen scavenger: reproducibility, accuracy, stability, and dose rate dependence" [Med. Phys. 33, 2506-2518 (2006)].
PMID 17278522