EMAIL THIS PAGE TO A FRIEND
Future oncology (London, England)
Future roles for FoxM1 inhibitors in cancer treatments.
PMID 17280494
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S6076
Siomycin A, from Streptomyces sioyaensis, ≥98% (HPLC)
C71H81N19O18S5