EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Chiral oxazaborolidine-aluminum bromide complexes are unusually powerful and effective catalysts for enantioselective Diels-Alder reactions.
PMID 17283985
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
399612
Aluminum diboride, −325 mesh
AlB2