EMAIL THIS PAGE TO A FRIEND
Organic letters
Gold(III)-catalyzed double hydroamination of o-alkynylaniline with terminal alkynes leading to N-vinylindoles.
PMID 17286370
Abstract
A highly efficient double-hydroamination reaction of o-alkynylanilines with terminal alkynes leading to N-alkenylindoles was developed by using gold(III) as a catalyst under neat conditions. [reaction: see text].
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
206490
4-Ethynylanisole, 97%
C9H8O