EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Syntheses of novel chiral sulfinamido ligands and their applications in diethylzinc additions to aldehydes.
PMID 17288383
Abstract
A novel class of sulfinamido alcohol ligands 1-6 was synthesized from (S)-tert-butanesulfinamide. These ligands showed excellent catalytic activities and enantiomeric selectivities in asymmetric additions of diethylzinc to aromatic aldehydes.