EMAIL THIS PAGE TO A FRIEND
Endocrinology
A role for beta-cell-expressed G protein-coupled receptor 119 in glycemic control by enhancing glucose-dependent insulin release.
PMID 17289847
Abstract
Pancreatic beta-cell dysfunction is a hallmark event in the pathogenesis of type 2 diabetes. Injectable peptide agonists of the glucagon-like peptide 1 (GLP-1) receptor have shown significant promise as antidiabetic agents by virtue of their ability to amplify glucose-dependent insulin release and preserve pancreatic beta-cell mass. These effects are mediated via stimulation of cAMP through beta-cell GLP-1 receptors. We report that the Galpha(s)-coupled receptor GPR119 is largely restricted to insulin-producing beta-cells of pancreatic islets. Additionally, we show here that GPR119 functions as a glucose-dependent insulinotropic receptor. Unlike receptors for GLP-1 and other peptides that mediate enhanced glucose-dependent insulin release, GPR119 was suitable for the development of potent, orally active, small-molecule agonists. The GPR119-specific agonist AR231453 significantly increased cAMP accumulation and insulin release in both HIT-T15 cells and rodent islets. In both cases, loss of GPR119 rendered AR231453 inactive. AR231453 also enhanced glucose-dependent insulin release in vivo and improved oral glucose tolerance in wild-type mice but not in GPR119-deficient mice. Diabetic KK/A(y) mice were also highly responsive to AR231453. Orally active GPR119 agonists may offer significant promise as novel antihyperglycemic agents acting in a glucose-dependent fashion.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML1883
AR231453, ≥98% (HPLC)
C21H24FN7O5S