EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
EPR spectra of Cu(2+) doped [Zn(sac)2(dmen)] and [Zn(sac)2(paen)] single crystals.
PMID 17292662
Abstract
Cu(2+) doped single crystals of [Zn(sac)2(dmen)] (sac: saccharinate, dmen: N,N'-dimethylethylendiamine) and [Zn(sac)2(paen)], (paen: N,N'-bis(3-propylamine)ethylendiamine) complexes have been investigated by electron paramagnetic resonance (EPR) technique. Detailed investigations of the EPR spectra indicate that Cu(2+) ion substitute with Zn(2+) ion and forms tetrahedral complex in [Zn(sac)2(dmen)] and octahedral complex in [Zn(sac)2(paen)] hosts. Principal values of the g and hyperfine tensors are determined and the ground state wave functions of Cu(2+) ions are obtained using EPR parameters.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D157805
N,N′-Dimethylethylenediamine, 98%
C4H12N2
270032
N,N′-Dimethylethylenediamine, 85%
C4H12N2